Assay Details
Target Gene Details
Entrez Gene ID: | 408190 |
Gene Name: | WAP four-disulfide core domain 13 |
Gene Aliases: |
Wfdc13l1 |
Location: |
Chr.2:164685107-164687706 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wfdc13 | NM_001012704.1 | NP_001012722.1 | ||
| AY542486.1 | AAT27474.1 | |||
| AY542487.1 | AAT27475.1 |
More Information
Set Membership: |
|

Genomic Map