Assay Details
Target Gene Details
Entrez Gene ID: | 56791 |
Gene Name: | ubiquitin-conjugating enzyme E2L 6 |
Gene Aliases: |
2810489I21Rik, RIG-B, UBCH8, Ubce8, Ubcm8 |
Location: |
Chr.2:84798828-84810003 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2l6 | NM_019949.2 | NP_064333.2 | ||
| AF159230.1 | AAD55978.1 | |||
| AK010942.1 | BAB27282.1 | |||
| AK013452.1 | BAB28861.1 | |||
| AK152010.1 | BAE30873.1 | |||
| AK152066.1 | ||||
| AK152113.1 | BAE30957.1 | |||
| AK168695.1 | BAE40539.1 | |||
| AK171629.1 | BAE42574.1 | |||
| BC008238.1 | AAH08238.1 | |||
| CT010270.1 | CAJ18478.1 |
More Information
Set Membership: |
|

Genomic Map