Assay Details
Target Gene Details
Entrez Gene ID: | 58170 |
Gene Name: | acid-sensing (proton-gated) ion channel family member 5 |
Gene Aliases: |
Accn5, Blinac, Inac |
Location: |
Chr.3:81982293-82021233 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asic5 | NM_021370.2 | NP_067345.1 | ||
| AK038321.1 | ||||
| Y19035.1 | CAB54040.1 |
More Information
Set Membership: |
|

Genomic Map