Assay Details
Target Gene Details
Entrez Gene ID: | 56456 |
Gene Name: | actin-like 6A |
Gene Aliases: |
2810432C06Rik, AI851094, ARP4, Actl6, Baf53a, C79802 |
Location: |
Chr.3:32708546-32726971 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actl6a | NM_019673.2 | NP_062647.2 | ||
| AF041476.1 | AAC94992.1 | |||
| AK002724.1 | ||||
| AK090154.1 | BAC41116.1 | |||
| AK167905.1 | BAE39913.1 | |||
| AK169278.1 | BAE41036.1 | |||
| BC001994.1 | AAH01994.1 | |||
| BC094498.1 | AAH94498.1 |
More Information
Set Membership: |
|

Genomic Map