Assay Details
Target Gene Details
Entrez Gene ID: | 22138 |
Gene Name: | titin |
Gene Aliases: |
1100001C23Rik, 2310036G12Rik, 2310057K23Rik, 2310074I15Rik, AF006999, AV006427, D330041I19Rik, D830007G01Rik, L56, mdm, shru |
Location: |
Chr.2:76703980-76982557 on Build GRCm38 |
Assay Gene Location: | Within Exon 309 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttn | NM_011652.3 | 308 | 98986 | NP_035782.3 |
| NM_028004.2 | 187 | 79243 | NP_082280.2 | |
| AK029024.1 | 1 | 84 | BAC26249.1 |

Genomic Map