Assay Details
Target Gene Details
Entrez Gene ID: | 100503106 |
Gene Name: | predicted gene 13944 |
Gene Aliases: |
OTTMUSG00000015155 |
Location: |
Chr.2:77462644-77480938 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm13944 | NR_040368.1 | |||
AK078367.1 | ||||
CJ104421.1 |
Target Gene Details
Entrez Gene ID: | 241494 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
B830010L13Rik, C130013B13Rik, Zfp533, Znf385b |
Location: |
Chr.2:77410622-77817706 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp385b | NM_001113399.1 | NP_001106870.1 | ||
NM_001113400.1 | NP_001106871.1 | |||
NM_178723.6 | NP_848838.2 | |||
AK046798.1 | BAC32874.1 | |||
AK050293.1 | BAC34170.1 | |||
AK078551.1 | ||||
AK079322.1 | ||||
AK149352.1 | BAE28828.1 | |||
BC048543.1 | AAH48543.1 | |||
BC132352.1 | ||||
BC138239.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
