Assay Details
Target Gene Details
Entrez Gene ID: | 65111 |
Gene Name: | death associated protein 3 |
Gene Aliases: |
4921514D13Rik, DAP-3, MRP-S29, S29mt |
Location: |
Chr.3:88920803-88951266 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dap3 | NM_001164533.1 | NP_001158005.1 | ||
| NM_022994.3 | NP_075370.2 | |||
| AJ250375.1 | CAC05583.1 | |||
| AK014889.1 | BAB29605.1 | |||
| AK021250.1 | ||||
| AK032275.1 | BAC27790.1 | |||
| AK088960.1 | BAC40673.1 | |||
| BC019566.1 | AAH19566.2 | |||
| CD741520.1 |
More Information
Set Membership: |
|

Genomic Map