Assay Details
Target Gene Details
Entrez Gene ID: | 28030 |
Gene Name: | G elongation factor, mitochondrial 1 |
Gene Aliases: |
AW545374, D3Wsu133e, Gfm |
Location: |
Chr.3:67430115-67475068 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfm1 | NM_138591.2 | 4 | 587 | NP_613057.2 |
| AF315511.1 | 4 | 554 | AAK58878.1 | |
| AK033214.1 | 4 | 586 | ||
| AK215134.1 | ||||
| BC013093.1 | 4 | 590 | AAH13093.1 | |
| BC031772.1 | 4 | 554 | AAH31772.1 |

Genomic Map