Assay Details
Target Gene Details
Entrez Gene ID: | 50754 |
Gene Name: | F-box and WD-40 domain protein 7 |
Gene Aliases: |
1110001A17Rik, AGO, Cdc4, Fbw7, Fbwd6, Fbx30, Fbxo30, Fbxw6, SEL-10 |
Location: |
Chr.3:84815577-84979198 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbxw7 | NM_001177773.1 | NP_001171244.1 | ||
NM_001177774.1 | NP_001171245.1 | |||
NM_080428.3 | NP_536353.2 | |||
AB221593.1 | ||||
AF391192.1 | AAL40928.1 | |||
AF427101.1 | AAL50052.1 | |||
BC085184.1 | ||||
BC131648.1 |
