Assay Details
Target Gene Details
Entrez Gene ID: | 171388 |
Gene Name: | BCL2/adenovirus E1B 19kD interacting protein like |
Gene Aliases: |
1700128A13Rik, BNIP-S, BNIPL1, BNIPL2, PP73, PP753 |
Location: |
Chr.3:95241293-95251193 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bnipl | NM_001168356.1 | NP_001161828.1 | ||
NM_134253.2 | NP_599014.2 | |||
AK142472.1 | BAE25078.1 | |||
BC005659.1 | AAH05659.1 | |||
BC006585.1 | AAH06585.1 |
