Assay Details
Target Gene Details
Entrez Gene ID: | 70285 |
Gene Name: | ribosome production factor 1 homolog (S. cerevisiae) |
Gene Aliases: |
2210420E24Rik, 2310066N05Rik, Bxdc5 |
Location: |
Chr.3:146506344-146521431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpf1 | NM_027332.3 | NP_081608.1 | ||
| NM_027371.3 | NP_081647.2 | |||
| AK008988.1 | ||||
| AK010079.1 | ||||
| AK162073.1 | ||||
| AK171035.1 | ||||
| AK171079.1 | BAE42233.1 | |||
| BC056165.1 | ||||
| BC089369.1 |
More Information
Set Membership: |
|

Genomic Map