Assay Details
Target Gene Details
Entrez Gene ID: | 13809 |
Gene Name: | glutamyl aminopeptidase |
Gene Aliases: |
6030431M22Rik, APA, Bp-1/6C3, Ly-51, Ly51 |
Location: |
Chr.3:129269177-129332749 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enpep | NM_007934.3 | NP_031960.1 | ||
| AK031439.1 | BAC27405.1 | |||
| AK034613.1 | ||||
| AK128975.1 | ||||
| AK146808.1 | BAE27448.1 | |||
| BC064061.1 | AAH64061.1 | |||
| BC127060.1 | ||||
| DQ000292.1 | AAY16106.1 | |||
| M29961.1 | AAB47732.1 |
More Information
Set Membership: |
|

Genomic Map