Assay Details
Target Gene Details
Entrez Gene ID: | 68723 |
Gene Name: | hornerin |
Gene Aliases: |
1110033K19Rik, A530063N20Rik, S100a18 |
Location: |
Chr.3:93319541-93333572 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hrnr | NM_133698.2 | 2 | 124 | NP_598459.2 |
| AK041026.1 | 2 | 125 | BAC30787.1 | |
| BC118008.1 | 1 | 87 |
More Information
Set Membership: |
|

Genomic Map