Assay Details
Target Gene Details
Entrez Gene ID: | 67006 |
Gene Name: | CDGSH iron sulfur domain 2 |
Gene Aliases: |
1500009M05Rik, 1500026J14Rik, 1500031D15Rik, AI848398, B630006A20Rik, Miner1, Noxp70, Zcd2 |
Location: |
Chr.3:135406412-135424442 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cisd2 | NM_025902.3 | NP_080178.1 | ||
| AK003486.1 | BAB22814.1 | |||
| AK004257.1 | BAB23237.1 | |||
| AK005184.1 | BAB23868.1 | |||
| AK005301.1 | ||||
| AM162549.1 | CAJ44265.1 | |||
| BC058279.1 | AAH58279.1 |

Genomic Map