Assay Details
Target Gene Details
Entrez Gene ID: | 14686 |
Gene Name: | guanine nucleotide binding protein, alpha transducing 2 |
Gene Aliases: |
AW490837, Gnat-2, Gt-2, Tcalpha |
Location: |
Chr.3:108086804-108101432 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnat2 | NM_008141.3 | NP_032167.1 | ||
| AK009388.1 | BAB26258.2 | |||
| BC016272.1 | AAH16272.1 | |||
| BC131941.1 | ||||
| BC131943.1 | ||||
| L10666.1 | AAC37650.1 |
More Information
Set Membership: |
|

Genomic Map