Assay Details
Target Gene Details
Entrez Gene ID: | 73124 |
Gene Name: | golgi integral membrane protein 4 |
Gene Aliases: |
3110027H23Rik, GIMPC, GPP130, Golph4, P138 |
Location: |
Chr.3:75876183-75956949 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golim4 | NM_001291069.1 | NP_001277998.1 | ||
| NM_175193.6 | NP_780402.1 | |||
| AK033730.1 | ||||
| AK048434.1 | BAC33335.1 | |||
| AK078928.1 | ||||
| BC086642.1 | AAH86642.1 | |||
| BC089354.1 | AAH89354.1 |

Genomic Map