Assay Details
Target Gene Details
Entrez Gene ID: | 242248 |
Gene Name: | B cell scaffold protein with ankyrin repeats 1 |
Gene Aliases: |
A530094C12Rik, AI451642, AVIEF, BANK |
Location: |
Chr.3:136053363-136326141 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bank1 | NM_001033350.2 | NP_001028522.2 | ||
AK041071.1 | ||||
AK041242.1 | BAC30875.1 | |||
AK156201.1 | BAE33622.1 | |||
AY178734.1 | AAO13613.1 | |||
BC026932.1 | ||||
BC116328.1 | ||||
EU051378.1 |
