Assay Details
Target Gene Details
Entrez Gene ID: | 242248 |
Gene Name: | B cell scaffold protein with ankyrin repeats 1 |
Gene Aliases: |
A530094C12Rik, AI451642, AVIEF, BANK |
Location: |
Chr.3:136053363-136326141 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bank1 | NM_001033350.2 | 7 | 1214 | NP_001028522.2 |
AK041071.1 | 4 | 472 | ||
AK041242.1 | 7 | 1175 | BAC30875.1 | |
AK156201.1 | 7 | 1224 | BAE33622.1 | |
AY178734.1 | 7 | 1234 | AAO13613.1 | |
BC026932.1 | 7 | 1161 | ||
BC116328.1 | ||||
EU051378.1 | 6 | 815 |
