Assay Details
Target Gene Details
Entrez Gene ID: | 71492 |
Gene Name: | Bardet-Biedl syndrome 7 (human) |
Gene Aliases: |
8430406N16Rik |
Location: |
Chr.3:36573140-36613489 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bbs7 | NM_027810.3 | 4 | 351 | NP_082086.2 |
AF521645.1 | 4 | 350 | AAO16027.1 | |
AK018389.1 | 4 | 350 | BAB31190.1 | |
AK050291.1 | 4 | 349 | BAC34169.1 | |
AK132661.1 | 4 | 279 | BAE21287.1 | |
BC031505.1 | 4 | 301 | AAH31505.1 |
