Assay Details
Target Gene Details
Entrez Gene ID: | 68259 |
Gene Name: | intraflagellar transport 80 |
Gene Aliases: |
4921524P20Rik, Wdr56, mKIAA1374 |
Location: |
Chr.3:68892499-69004653 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift80 | NM_026641.2 | NP_080917.1 | ||
| AK042231.1 | BAC31200.1 | |||
| AK129339.1 | BAC98149.1 | |||
| AK147305.1 | ||||
| BC013814.1 | AAH13814.1 | |||
| BC034101.1 | AAH34101.1 |

Genomic Map