Assay Details
Target Gene Details
Entrez Gene ID: | 12350 |
Gene Name: | carbonic anhydrase 3 |
Gene Aliases: |
BB219044, Ca3, Car-3 |
Location: |
Chr.3:14863490-14872536 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car3 | NM_007606.3 | 2 | 204 | NP_031632.2 |
| AK002349.1 | 2 | 231 | ||
| AK003671.1 | 2 | 234 | ||
| AK017227.1 | 2 | 231 | ||
| AK075630.1 | 1 | 878 | ||
| BC011129.1 | 2 | 204 | AAH11129.1 | |
| M27796.1 | 2 | 253 | AAA37355.1 |

Genomic Map