Assay Details
Target Gene Details
Entrez Gene ID: | 14466 |
Gene Name: | glucosidase, beta, acid |
Gene Aliases: |
GBA1, GC, GCase, GLUC, betaGC |
Location: |
Chr.3:89202914-89208966 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gba | NM_001077411.2 | 8 | 1230 | NP_001070879.1 |
| NM_008094.5 | 9 | 1399 | NP_032120.1 | |
| NR_122037.1 | 9 | 1399 | ||
| AK082767.1 | 8 | 1228 | BAC38609.1 | |
| AK152745.1 | 8 | 1211 | ||
| AK166198.1 | 9 | 1390 | ||
| AK205177.1 | 1 | 88 | ||
| BC006663.1 | 9 | 1384 | AAH06663.1 | |
| M24119.1 | 8 | 1139 | AAA37671.1 |

Genomic Map