Assay Details
Target Gene Details
Entrez Gene ID: | 229841 |
Gene Name: | centromere protein E |
Gene Aliases: |
312kDa, AU019344, BC049989, C530022J18, CENP-E, Kif10 |
Location: |
Chr.3:135212563-135273540 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cenpe | NM_173762.4 | NP_776123.3 | ||
| AY493378.1 | AAR85498.1 |

Genomic Map