Assay Details
Target Gene Details
Entrez Gene ID: | 13123 |
Gene Name: | cytochrome P450, family 7, subfamily b, polypeptide 1 |
Gene Aliases: |
AW261589, D3Ertd552e, hct-1 |
Location: |
Chr.3:18071950-18243893 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp7b1 | NM_007825.4 | NP_031851.3 | ||
| AK013034.1 | BAB28613.1 | |||
| AK140100.1 | BAE24236.1 | |||
| AK172152.1 | BAE42849.1 | |||
| BC038810.1 | AAH38810.1 | |||
| CJ065666.1 | ||||
| U36993.1 | AAA92615.1 |
More Information
Set Membership: |
|

Genomic Map