Assay Details
Target Gene Details
Entrez Gene ID: | 72413 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 2 |
Gene Aliases: |
2700049B16Rik, 3110031N04Rik |
Location: |
Chr.3:31902132-32200180 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnmb2 | NM_028231.2 | 4 | 488 | NP_082507.1 |
| AB193520.1 | 2 | 98 | ||
| AB193521.1 | ||||
| AK012400.1 | 4 | 339 | BAB28216.1 | |
| AK014106.1 | 3 | 585 | ||
| AY062429.1 | 2 | 98 | AAL38982.1 | |
| BC046227.1 | 4 | 488 | AAH46227.1 | |
| BC058957.1 | 4 | 488 | AAH58957.1 |
More Information
Set Membership: |
|

Genomic Map