Assay Details
Target Gene Details
Entrez Gene ID: | 100502876 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 3 |
Gene Aliases: |
EG435726, Gm5707 |
Location: |
Chr.3:32471614-32502807 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnmb3 | NM_001195074.1 | NP_001182003.1 |
More Information
Set Membership: |
|

Genomic Map