Assay Details
Target Gene Details
Entrez Gene ID: | 66815 |
Gene Name: | coiled-coil domain containing 109B |
Gene Aliases: |
9030408N13Rik, MCUb |
Location: |
Chr.3:129914960-129970214 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc109b | NM_001293644.1 | 7 | 999 | NP_001280573.1 |
| NM_025779.3 | 8 | 1160 | NP_080055.2 | |
| AK013032.1 | 4 | 468 | BAB28611.1 | |
| AK018492.1 | 8 | 1037 | BAB31237.1 | |
| AK150724.1 | 8 | 1058 | BAE29802.1 | |
| AK150995.1 | 8 | 1034 | BAE30019.1 | |
| AK151201.1 | 8 | 1058 | BAE30198.1 | |
| AK151915.1 | 8 | 1071 | BAE30794.1 | |
| AK152326.1 | 7 | 870 | ||
| AK154338.1 | 8 | 1053 | BAE32524.1 | |
| BC049571.1 | 8 | 1160 | AAH49571.1 | |
| BX515874.1 | 3 | 211 |

Genomic Map