Assay Details
Target Gene Details
Entrez Gene ID: | 12630 |
Gene Name: | complement component factor i |
Gene Aliases: |
- |
Location: |
Chr.3:129835889-129875329 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfi | NM_007686.2 | 1 | 64 | NP_031712.2 |
| BC150751.1 | 1 | 57 | ||
| BY773461.1 | 1 | 65 | ||
| CJ133989.1 | ||||
| U47810.1 | 1 | 67 | AAB00438.1 |
More Information
Set Membership: |
|

Genomic Map