Assay Details
Target Gene Details
Entrez Gene ID: | 229672 |
Gene Name: | BCLl2-like 15 |
Gene Aliases: |
Bfk, Gm566 |
Location: |
Chr.3:103831039-103854618 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l15 | NM_001142959.1 | 3 | 511 | NP_001136431.1 |
| NM_001142960.1 | 3 | 511 | NP_001136432.1 | |
| AF014453.1 | 3 | 607 | ||
| BC027668.1 | 3 | 347 | AAH27668.1 | |
| BC117780.1 | 3 | 511 | ||
| BC117781.1 | 3 | 511 | ||
| BC128505.1 | 3 | 511 | ||
| BC128506.1 | 3 | 511 |

Genomic Map