Assay Details
Target Gene Details
Entrez Gene ID: | 12630 |
Gene Name: | complement component factor i |
Gene Aliases: |
- |
Location: |
Chr.3:129835889-129875329 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfi | NM_007686.2 | NP_031712.2 | ||
| BC150751.1 | ||||
| CN691735.1 | ||||
| U47810.1 | AAB00438.1 |
More Information
Set Membership: |
|

Genomic Map