Assay Details
Target Gene Details
Entrez Gene ID: | 66326 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 5 beta |
Gene Aliases: |
1700008A05Rik |
Location: |
Chr.3:19508591-19610862 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc5b | NM_001163536.1 | NP_001157008.1 | ||
| NM_001163537.1 | NP_001157009.1 | |||
| NM_025489.3 | NP_079765.3 | |||
| AK005725.1 | BAB24206.1 | |||
| AK005749.1 | BAB24221.1 | |||
| BC049579.1 | AAH49579.1 | |||
| CA463124.1 |
More Information
Set Membership: |
|

Genomic Map