Assay Details
Target Gene Details
Entrez Gene ID: | 67553 |
Gene Name: | glutathione S-transferase, C-terminal domain containing |
Gene Aliases: |
4933434L15Rik |
Location: |
Chr.3:132981752-133091971 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gstcd | NM_026231.2 | NP_080507.2 | ||
AK013967.1 | BAB29091.1 | |||
AK017059.1 | BAB30574.2 | |||
AK020021.1 | BAB31969.1 | |||
AK029724.1 | ||||
AK033409.1 | ||||
BC047136.1 | AAH47136.1 | |||
BC061240.1 | AAH61240.1 |
