Assay Details
Target Gene Details
Entrez Gene ID: | 14862 |
Gene Name: | glutathione S-transferase, mu 1 |
Gene Aliases: |
Gstb-1, Gstb1 |
Location: |
Chr.3:108012250-108017973 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstm1 | NM_010358.5 | 1 | 72 | NP_034488.1 |
| J04632.1 | 1 | 73 | AAA37705.1 |
More Information
Set Membership: |
|

Genomic Map