Assay Details
Target Gene Details
Entrez Gene ID: | 69772 |
Gene Name: | 3-hydroxybutyrate dehydrogenase, type 2 |
Gene Aliases: |
1810026B04Rik, Dhrs6 |
Location: |
Chr.3:135281221-135304426 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bdh2 | NM_001172055.1 | NP_001165526.1 | ||
| NM_027208.2 | NP_081484.1 | |||
| AK007603.1 | ||||
| BC036998.1 | AAH36998.1 |
More Information
Set Membership: |
|

Genomic Map