Assay Details
Target Gene Details
Entrez Gene ID: | 67707 |
Gene Name: | mitochondrial ribosomal protein L24 |
Gene Aliases: |
2010005E08Rik, 2810470K06Rik, 6720473G22Rik, AA407670 |
Location: |
Chr.3:87919074-87923434 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl24 | NM_026591.3 | NP_080867.2 | ||
| AK003362.1 | BAB22737.1 | |||
| AK013394.1 | BAB28828.1 | |||
| AK016190.1 | ||||
| AK019976.1 | BAB31945.1 | |||
| AK020157.1 | BAB32014.1 | |||
| AK167623.1 | BAE39675.1 | |||
| BC025506.1 | AAH25506.1 | |||
| BC031730.1 | AAH31730.1 |
More Information
Set Membership: |
|

Genomic Map