Assay Details
Target Gene Details
Entrez Gene ID: | 74600 |
Gene Name: | mitochondrial ribosomal protein L47 |
Gene Aliases: |
4833424P18Rik, CGI-204, ENSMUSG00000051761, Gm9859, L47mt, MRP-L47, MTF/L47, NCM1 |
Location: |
Chr.3:32727497-32736755 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl47 | NM_029017.2 | NP_083293.1 | ||
| AA174956.1 | ||||
| BC038065.1 | AAH38065.1 | |||
| BC087953.1 | AAH87953.1 |
More Information
Set Membership: |
|

Genomic Map