Assay Details
Target Gene Details
Entrez Gene ID: | 53814 |
Gene Name: | ornithine decarboxylase antizyme 3 |
Gene Aliases: |
AZ3, Az-3, Oaz-t |
Location: |
Chr.3:94433387-94436651 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oaz3 | NM_016901.3 | 4 | 653 | NP_058597.2 |
| AB045835.1 | 4 | 578 | BAB97207.1 | |
| AF175297.1 | 4 | 618 | AAD51735.1 | |
| AI449747.1 | 2 | 303 | ||
| AK006116.1 | 4 | 588 | ||
| AK006634.1 | 4 | 589 | ||
| AK076942.1 | 4 | 595 |
More Information
Set Membership: |
|

Genomic Map