Assay Details
Target Gene Details
Entrez Gene ID: | 229487 |
Gene Name: | glutamyl-tRNA(Gln) amidotransferase, subunit B |
Gene Aliases: |
9430026F02Rik, Pet112, Pet112l |
Location: |
Chr.3:85574102-85655660 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gatb | NM_144896.4 | NP_659145.1 | ||
| AK028193.1 | BAC25803.1 | |||
| AK034702.1 | BAC28801.1 | |||
| AK053978.1 | BAC35607.1 | |||
| AK081800.1 | BAC38336.1 | |||
| AK134831.1 | ||||
| AK166231.1 | BAE38646.1 | |||
| AK182374.1 | ||||
| BC005709.1 | AAH05709.1 |
More Information
Set Membership: |
|

Genomic Map