Assay Details
Target Gene Details
Entrez Gene ID: | 56692 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 3 |
Gene Aliases: |
AW556229, Map2k1ip1, Mapbp, Mapksp1, Mp1 |
Location: |
Chr.3:137918555-137928762 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamtor3 | NM_019920.2 | NP_064304.1 | ||
| AF082526.1 | AAC34591.1 | |||
| AK031303.1 | 1 | 1169 | ||
| AK088454.1 | BAC40361.1 | |||
| AK151432.1 | BAE30395.1 | |||
| AK159450.1 | BAE35093.1 | |||
| AK162102.1 | BAE36726.1 | |||
| BC016417.1 | AAH16417.1 | |||
| BC083155.1 | AAH83155.1 |
More Information
Set Membership: |
|

Genomic Map