Assay Details
Target Gene Details
Entrez Gene ID: | 97112 |
Gene Name: | NMD3 homolog (S. cerevisiae) |
Gene Aliases: |
C87860 |
Location: |
Chr.3:69722055-69749047 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nmd3 | NM_133787.2 | NP_598548.1 | ||
AK088667.1 | BAC40491.1 | |||
AK143289.1 | BAE25334.1 | |||
AK145248.1 | BAE26326.1 | |||
AK157890.1 | 1 | 1400 | BAE34248.1 | |
AK166283.1 | BAE38680.1 | |||
BC003842.1 | AAH03842.1 |
