Assay Details
Target Gene Details
Entrez Gene ID: | 18805 |
Gene Name: | phospholipase D1 |
Gene Aliases: |
AA536939, C85393, Pld1a, Pld1b |
Location: |
Chr.3:27938680-28133362 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pld1 | NM_001164056.1 | 16 | 1935 | NP_001157528.1 |
| NM_008875.4 | 15 | 1880 | NP_032901.2 | |
| AK136111.1 | 10 | 1118 | BAE22827.1 | |
| AK149229.1 | ||||
| BC068144.1 | 15 | 1737 | AAH68144.1 | |
| U87868.1 | 16 | 1918 | AAB81245.1 |
More Information
Set Membership: |
|

Genomic Map