Assay Details
Target Gene Details
Entrez Gene ID: | 71325 |
Gene Name: | trichohyalin-like 1 |
Gene Aliases: |
5430400H23Rik, AA589420, S100a17, Thhl1 |
Location: |
Chr.3:93468734-93471980 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tchhl1 | NM_027762.3 | NP_082038.2 | ||
AK017244.1 | BAB30648.1 |
