Assay Details
Target Gene Details
Entrez Gene ID: | 56720 |
Gene Name: | tryptophan 2,3-dioxygenase |
Gene Aliases: |
AA407491, TDO, TO, chky |
Location: |
Chr.3:81958412-81975728 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdo2 | NM_019911.2 | NP_064295.2 | ||
| AB476989.1 | ||||
| AB476990.1 | ||||
| AK132271.1 | ||||
| BC018390.1 | AAH18390.1 | |||
| S56368.1 | ||||
| U24493.1 | AAB60491.1 |
More Information
Set Membership: |
|

Genomic Map