Assay Details
Target Gene Details
Entrez Gene ID: | 67472 |
Gene Name: | mitochondrial fission regulator 1 |
Gene Aliases: |
1300002C08Rik, 1700080D04Rik, 2810026O10Rik, 4930579E05Rik, Chppr, Fam54a2 |
Location: |
Chr.3:19187326-19220817 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtfr1 | NM_001253390.1 | NP_001240319.1 | ||
| NM_001253391.1 | NP_001240320.1 | |||
| NM_026182.5 | NP_080458.1 | |||
| NR_045575.1 | ||||
| AF354708.1 | AAK26434.1 | |||
| AK004846.1 | BAB23612.2 | |||
| AK006952.1 | BAB24802.2 | |||
| AK009195.2 | ||||
| AK016316.1 | ||||
| AK018048.1 | ||||
| AK075822.1 | BAC35988.1 | |||
| AK075992.1 | BAC36103.1 | |||
| AK134822.1 | ||||
| AK161251.1 | BAE36270.1 | |||
| AK193362.1 | ||||
| BC019971.1 | AAH19971.1 | |||
| BC042471.1 | AAH42471.1 | |||
| CK625955.1 | ||||
| DQ369744.1 | ABC96324.1 |

Genomic Map