Assay Details
Target Gene Details
Entrez Gene ID: | 67472 |
Gene Name: | mitochondrial fission regulator 1 |
Gene Aliases: |
1300002C08Rik, 1700080D04Rik, 2810026O10Rik, 4930579E05Rik, Chppr, Fam54a2 |
Location: |
Chr.3:19187326-19220817 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtfr1 | NM_001253390.1 | 5 | 531 | NP_001240319.1 |
NM_001253391.1 | 5 | 693 | NP_001240320.1 | |
NM_026182.5 | 5 | 541 | NP_080458.1 | |
NR_045575.1 | 6 | 578 | ||
AF354708.1 | 4 | 492 | AAK26434.1 | |
AK004846.1 | 5 | 519 | BAB23612.2 | |
AK006952.1 | 5 | 693 | BAB24802.2 | |
AK009195.2 | 5 | 507 | ||
AK016316.1 | 5 | 680 | ||
AK018048.1 | 5 | 523 | ||
AK075822.1 | 5 | 517 | BAC35988.1 | |
AK075992.1 | 5 | 541 | BAC36103.1 | |
AK134822.1 | 6 | 535 | ||
AK161251.1 | 5 | 514 | BAE36270.1 | |
AK193362.1 | 2 | 183 | ||
BC019971.1 | 5 | 489 | AAH19971.1 | |
BC042471.1 | 4 | 465 | AAH42471.1 | |
CK625955.1 | 5 | 521 | ||
DQ369744.1 | 5 | 652 | ABC96324.1 |
