Assay Details
Target Gene Details
Entrez Gene ID: | 67472 |
Gene Name: | mitochondrial fission regulator 1 |
Gene Aliases: |
1300002C08Rik, 1700080D04Rik, 2810026O10Rik, 4930579E05Rik, Chppr, Fam54a2 |
Location: |
Chr.3:19187326-19220817 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtfr1 | NM_001253390.1 | 6 | 775 | NP_001240319.1 |
NM_001253391.1 | 6 | 937 | NP_001240320.1 | |
NM_026182.5 | 6 | 785 | NP_080458.1 | |
NR_045575.1 | 7 | 822 | ||
AF354708.1 | 5 | 736 | AAK26434.1 | |
AK004846.1 | 6 | 763 | BAB23612.2 | |
AK006952.1 | 6 | 937 | BAB24802.2 | |
AK009195.2 | 6 | 751 | ||
AK016316.1 | 6 | 925 | ||
AK018048.1 | 6 | 767 | ||
AK075822.1 | 6 | 761 | BAC35988.1 | |
AK075992.1 | 6 | 785 | BAC36103.1 | |
AK134822.1 | 7 | 779 | ||
AK161251.1 | 6 | 758 | BAE36270.1 | |
BC019971.1 | 6 | 733 | AAH19971.1 | |
BC042471.1 | 5 | 709 | AAH42471.1 | |
DQ369744.1 | 6 | 896 | ABC96324.1 |
