Assay Details
Target Gene Details
Entrez Gene ID: | 242202 |
Gene Name: | phosphodiesterase 5A, cGMP-specific |
Gene Aliases: |
Cgbpde, Cn5n, Pde5, Pde5a1 |
Location: |
Chr.3:122729158-122859374 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde5a | NM_153422.2 | 10 | 1538 | NP_700471.2 |
| AF541937.1 | 10 | 1538 | AAN17330.1 | |
| BC119137.1 | 10 | 1501 | ||
| BC120486.1 | 10 | 1501 |
More Information
Set Membership: |
|

Genomic Map