Assay Details
Target Gene Details
Entrez Gene ID: | 20202 |
Gene Name: | S100 calcium binding protein A9 (calgranulin B) |
Gene Aliases: |
60B8Ag, AW546964, BEE22, Cagb, GAGB, L1Ag, MRP14, p14 |
Location: |
Chr.3:90692630-90695721 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a9 | NM_001281852.1 | NP_001268781.1 | ||
| NM_009114.3 | NP_033140.1 | |||
| AK143826.1 | BAE25555.1 | |||
| BC027635.1 | AAH27635.1 | |||
| BY112406.1 | ||||
| M83219.1 | AAB07228.1 |
More Information
Set Membership: |
|

Genomic Map