Assay Details
Target Gene Details
Entrez Gene ID: | 140918 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 12 |
Gene Aliases: |
Asc-2, XAT1 |
Location: |
Chr.3:14480691-14505819 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a12 | NM_080852.2 | 2 | 832 | NP_543128.1 |
| AB055005.1 | 2 | 837 | BAB83620.1 | |
| AJ417663.1 | 2 | 794 | CAD10395.1 |
More Information
Set Membership: |
|

Genomic Map