Assay Details
Target Gene Details
Entrez Gene ID: | 18160 |
Gene Name: | natriuretic peptide receptor 1 |
Gene Aliases: |
AI893888, GC-A, NPR-A, NPRA, Pndr |
Location: |
Chr.3:90450592-90465928 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npr1 | NM_008727.5 | NP_032753.5 | ||
| AK135008.1 | BAE22383.1 | |||
| BC110659.1 | AAI10660.1 | |||
| J05504.1 | AAA37670.1 | |||
| L31932.1 | AAA66945.1 |
More Information
Set Membership: |
|

Genomic Map